(R)-(+)-N,α-Dimethylbenzylamin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(R)-N-Methyl-1-phenylethanamine, (R)-N-Methyl-alpha-phenylethylamine
394009-1G
394009-5G
SIAL394009-1GEA
118
EUR
SIAL394009-1G
SIAL394009-5G
(R)-(+)-N,α-Dimethylbenzylamin, Sigma-Aldrich®
(R)-(+)-N,α-Dimethylbenzylamin
Formel:
C₉H₁₃N Molecular Weight: 135,2062 g/mol |
MDL:
MFCD00044966 CAS-Nummer: 5933-40-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...