2-(2-Benzoylphenyl)-2-phenylacetonitril

Lieferant: BIONET RESEARCH

6G-040.500 6G-040.1 6G-040.5 6G-040.10
BIOT6G-040.500EA 0 EUR
BIOT6G-040.500 BIOT6G-040.1 BIOT6G-040.5 BIOT6G-040.10
2-(2-Benzoylphenyl)-2-phenylacetonitril
2-(2-Benzoylphenyl)-2-phenylacetonitril
Formel: C₂₁H₁₅NO
MDL: MFCD00231976
CAS-Nummer: 127667-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR