2-(4-Benzoylphenyl)-2-phenylacetonitril

Lieferant: BIONET RESEARCH

6G-043.500 6G-043.1 6G-043.5 6G-043.10
BIOT6G-043.500EA 0 EUR
BIOT6G-043.500 BIOT6G-043.1 BIOT6G-043.5 BIOT6G-043.10
2-(4-Benzoylphenyl)-2-phenylacetonitril
2-(4-Benzoylphenyl)-2-phenylacetonitril
Formel: C₂₁H₁₅NO
MDL: MFCD00231979
CAS-Nummer: 338791-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR