3,4-Methylendioxyphenylisothiocyanat

Lieferant: FLUOROCHEM

Synonyme: 1,3-Benzodioxol-5-yl isothiocyanate

009530-1G 009530-5G
FLUO009530-1GEA 31.1 EUR
FLUO009530-1G FLUO009530-5G
3,4-Methylendioxyphenylisothiocyanat
3,4-Methylendioxyphenylisothiocyanat
Formel: C₈H₅NO₂S
MDL: MFCD00066321
CAS-Nummer: 113504-93-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR