Chromon-6-boronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-8630-5G PN-8630-1G
COBBPN-8630-5GEA 1220 EUR
COBBPN-8630-5G COBBPN-8630-1G
Chromon-6-boronsäurepinakolester
Chromon-6-boronsäurepinakolester
Formel: C₁₅H₁₇BO₄
MDL: MFCD04038748
CAS-Nummer: 928773-42-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR