2-Amino-3-formylchromon
Lieferant: CHEMPUR
Synonyme:
2-Amino-4-oxo-4H-chromene-3-carbaldehyde
FL12025.POR
012025-5G
CHMPFL12025.POREA
0
EUR
CHMPFL12025.POR
FLUO012025-5G
2-Amino-3-formylchromon
2-Amino-3-formylchromon
Formel:
C₁₀H₇NO₃ Molecular Weight: 189,1675 g/mol |
MDL:
MFCD00191735 CAS-Nummer: 61424-76-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...