Furfuryl-methyldisulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-((Methyldisulphanyl)methyl)furan, Methyl furfuryl disulphide

M-Clarity™ Qualitätsstufe = MQ300

W336203-100G-K W336203-1KG-K
SIALW336203-100G-KEA 253 EUR
SIALW336203-100G-K SIALW336203-1KG-K
Furfuryl-methyldisulfid, Sigma-Aldrich®
Furfuryl-methyldisulfid
Formel: C₆H₈OS₂
Molecular Weight: 160,2571 g/mol
MDL: MFCD00012326
CAS-Nummer: 57500-00-2
EINECS: 260-773-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR