4-Indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-2411-5G PN-2411-1G
COBBPN-2411-5GEA 312 EUR
COBBPN-2411-5G COBBPN-2411-1G
4-Indolboronsäurepinakolester
4-Indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243.11 g/mol
MDL: MFCD08689896
CAS-Nummer: 388116-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR