5-Indolboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
Indole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
010834-5G
010834-1G
FLUO010834-5GEA
386
EUR
FLUO010834-5G
FLUO010834-1G
5-Indolboronsäurepinakolester
5-Indolboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₂ Molecular Weight: 243,1092 g/mol |
MDL:
MFCD03789263 CAS-Nummer: 269410-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...