5-Indolboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Indole-5-boronic acid pinacol ester, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

M-Clarity™ Qualitätsstufe = MQ100

578835-1G 578835-5G
SIAL578835-1GEA 49.8 EUR
SIAL578835-1G SIAL578835-5G
5-Indolboronsäurepinakolester, Sigma-Aldrich®
5-Indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243,1092 g/mol
MDL: MFCD03789263
CAS-Nummer: 269410-24-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR