6-Indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-2932-1G PN-2932-5G
COBBPN-2932-1GEA 137 EUR
COBBPN-2932-1G COBBPN-2932-5G
6-Indolboronsäurepinakolester
6-Indolboronsäurepinakolester
Formel: C₁₄H₁₈BNO₂
Molecular Weight: 243.1092 g/mol
MDL: MFCD07368852
CAS-Nummer: 642494-36-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR