7-Indolboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole
PN-8907-5G
PN-8907-1G
COBBPN-8907-5GEA
709
EUR
COBBPN-8907-5G
COBBPN-8907-1G
7-Indolboronsäurepinakolester
7-Indolboronsäurepinakolester
Formel:
C₁₄H₁₈BNO₂ Molecular Weight: 243.1092 g/mol |
MDL:
MFCD08669591 CAS-Nummer: 642494-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...