1-(Phenylsulfonyl)-5-indolboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 1-(Phenylsulphonyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

PN-5742-5G PN-5742-1G
COBBPN-5742-5GEA 1070 EUR
COBBPN-5742-5G COBBPN-5742-1G
1-(Phenylsulfonyl)-5-indolboronsäurepinakolester
1-(Phenylsulfonyl)-5-indolboronsäurepinakolester
Formel: C₂₀H₂₂BNO₄S
Molecular Weight: 383.2700 g/mol
MDL: MFCD12828206
CAS-Nummer: 1256359-23-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR