4-Indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Indolylboronic acid, (1H-Indol-4-yl)boronic acid

BB-2411-5G BB-2411-1G
COBBBB-2411-5GEA 625 EUR
COBBBB-2411-5G COBBBB-2411-1G
4-Indolboronsäure
4-Indoleboronic acid
Formel: C₈H₈BNO₂
Molecular Weight: 160.97 g/mol
MDL: MFCD03095175
CAS-Nummer: 220465-43-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR