1-(Phenylsulfonyl)-2-indolboronsäure
Lieferant: COMBI-BLOCKS
BB-8362-5G
BB-8362-1G
COBBBB-8362-5GEA
670
EUR
COBBBB-8362-5G
COBBBB-8362-1G
1-(Phenylsulfonyl)-2-indolboronsäure
1-(Phenylsulphonyl)-2-indoleboronic acid
Formel:
C₁₄H₁₂BNO₄S |
MDL:
MFCD03086094 CAS-Nummer: 342404-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...