1-(Phenylsulfonyl)-3-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(Phenylsulphonyl)-1H-indol-3-yl)boronic acid

BB-8357-5G BB-8357-1G
COBBBB-8357-5GEA 658 EUR
COBBBB-8357-5G COBBBB-8357-1G
1-(Phenylsulfonyl)-3-indolboronsäure
1-(Phenylsulphonyl)-3-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Molecular Weight: 301.1300 g/mol
MDL: MFCD02681892
CAS-Nummer: 129271-98-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR