1-(Phenylsulfonyl)-5-indolboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (1-(Phenylsulphonyl)-1H-indol-5-yl)boronic acid

BB-5742-5G BB-5742-1G
COBBBB-5742-5GEA 656 EUR
COBBBB-5742-5G COBBBB-5742-1G
1-(Phenylsulfonyl)-5-indolboronsäure
1-(Phenylsulphonyl)-5-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
Molecular Weight: 301.1300 g/mol
MDL: MFCD03840606
CAS-Nummer: 480438-51-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR