4-(Aminomethyl)tetrahydropyran HCl
Lieferant: COMBI-BLOCKS
Synonyme:
4-Aminomethyltetrahydropyran, HCl, 4-Aminomethyltetrahydropyran hydrochloride
AM-1013-5G
AM-1013-25G
AM-1013-1G
COBBAM-1013-5GEA
75.9
EUR
COBBAM-1013-5G
COBBAM-1013-25G
COBBAM-1013-1G
4-(Aminomethyl)tetrahydropyran HCl
4-(Aminomethyl)tetrahydropyran HCl
Formel:
C₆H₁₄ClNO Molecular Weight: 151,6300 g/mol |
MDL:
MFCD03411928 CAS-Nummer: 389621-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...