Tetrahydropyran-4-methanol
Lieferant: COMBI-BLOCKS
Synonyme:
4-(Hydroxymethyl)tetrahydropyran
AM-1017-5G
AM-1017-1G
COBBAM-1017-5GEA
63.9
EUR
COBBAM-1017-5G
COBBAM-1017-1G
Tetrahydropyran-4-methanol
Tetrahydropyran-4-methanol
Formel:
C₆H₁₂O₂ Molecular Weight: 116.16 g/mol |
MDL:
MFCD00457804 CAS-Nummer: 14774-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...