Tetrahydropyran-4-methanol

Lieferant: CHEMPUR

MO00962.250 MO00962.1
CHMPMO00962.250EA 0 EUR
CHMPMO00962.250 CHMPMO00962.1
Tetrahydropyran-4-methanol
Tetrahydropyran-4-methanol
Formel: C₆H₁₂O₂
MDL: MFCD00457804
CAS-Nummer: 14774-37-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR