3,4,5-Tribrom-1H-pyrazol
Lieferant: Thermo Scientific
Synonyme:
3,4,5-Tribromopyrazole
BTB06439.1
BTB06439.10
BTB06439.50
MAYBBTB06439.1EA
0
EUR
MAYBBTB06439.1
MAYBBTB06439.10
MAYBBTB06439.50
3,4,5-Tribrom-1H-pyrazol
3,4,5-Tribromo-1H-pyrazole
Formel:
C₃HBr₃N₂ Molecular Weight: 304,78 g/mol |
MDL:
MFCD00040248 CAS-Nummer: 17635-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...