1,3-Dimethyl-1H-pyrazol-5-carbonsäure
Lieferant: FLUOROCHEM
CFF457.2.5
CFF457.5
CFF457.10
017470-2.5G
017470-5G
017470-10G
CHMPCFF457.2.5EA
321
EUR
CHMPCFF457.2.5
CHMPCFF457.5
CHMPCFF457.10
FLUO017470-2.5G
FLUO017470-5G
FLUO017470-10G
1,3-Dimethyl-1H-pyrazol-5-carbonsäure
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formel:
C₆H₈N₂O₂ Molecular Weight: 140,1400 g/mol |
MDL:
MFCD00159636 CAS-Nummer: 5744-56-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...