1,3-Dimethyl-1H-pyrazol-5-carbonsäure

Lieferant: Thermo Scientific

CC12501.250 CC12501DA
MAYBCC12501.250EA 0 EUR
MAYBCC12501.250 MAYBCC12501DA
1,3-Dimethyl-1H-pyrazol-5-carbonsäure
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formel: C₆H₈N₂O₂
Molecular Weight: 140,1400 g/mol
MDL: MFCD00159636
CAS-Nummer: 5744-56-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR