1,3-Dimethyl-1H-pyrazol-5-carbonylchlorid
Lieferant: Thermo Scientific
CC12502.0001
CC12502CB
CC12502DA
MAYBCC12502.0001EA
0
EUR
MAYBCC12502.0001
MAYBCC12502CB
MAYBCC12502DA
1,3-Dimethyl-1H-pyrazol-5-carbonylchlorid
1,3-Dimethyl-1H-pyrazol-5-carbonylchlorid
Formel:
C₆H₇ClN₂O |
MDL:
MFCD00833258 CAS-Nummer: 55458-67-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...