1,4-Thioxan
Lieferant: FLUOROCHEM
001450-25G
001450-100G
FLUO001450-25GEA
171
EUR
FLUO001450-25G
FLUO001450-100G
1,4-Thioxan
1,4-Thioxan
Formel:
C₄H₈OS |
MDL:
MFCD00006573 CAS-Nummer: 15980-15-1 EINECS: 240-117-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...