2-Mercapto-5-(trifluormethoxy)benzothiazol

Lieferant: COMBI-BLOCKS

HC-3052-5G HC-3052-1G
COBBHC-3052-5GEA 595 EUR
COBBHC-3052-5G COBBHC-3052-1G
2-Mercapto-5-(trifluormethoxy)benzothiazol
2-Mercapto-5-(trifluoromethoxy)benzothiazole
Formel: C₈H₄F₃NOS₂
MDL: MFCD13195694
CAS-Nummer: 155559-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR