Benzothiazol-2-ol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Hydroxybenzothiazole
A16648.06
A16648.14
A16648.06EA
38.6
EUR
A16648.06
A16648.14
Benzothiazol-2-ol ≥98%
Benzothiazol-2-ol
Purity: 98%
CAS No.: 934-34-9
Molecular Formula: C7H5NOS
Formula Weight: 151.18
Melting Point: 136-140°
MDL No.: MFCD00022868
Formel:
C₇H₅NOS Molecular Weight: 151,18 g/mol Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00022868 CAS-Nummer: 934-34-9 EINECS: 213-281-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...