Benzothiazol-2-ol

Lieferant: FLUOROCHEM

Synonyme: 2-Hydroxybenzothiazole

009676-5G 009676-25G
FLUO009676-5GEA 0 EUR
FLUO009676-5G FLUO009676-25G
Benzothiazol-2-ol
Benzothiazol-2-ol
Formel: C₇H₅NOS
Molecular Weight: 151,18 g/mol
MDL: MFCD00022868
CAS-Nummer: 934-34-9
EINECS: 213-281-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR