Benzothiazol-2-ylmethanol

Lieferant: Thermo Scientific

Synonyme: 2-(Hydroxymethyl)benzothiazole, 2-Benzothiazolemethanol, 2-Hydroxymethylbenzothiazole

CC06009.250 CC06009.1
MAYBCC06009.250EA 0 EUR
MAYBCC06009.250 MAYBCC06009.1
Benzothiazol-2-ylmethanol
Benzothiazol-2-ylmethanol
Formel: C₈H₇NOS
Molecular Weight: 165.21 g/mol
MDL: MFCD00226293
CAS-Nummer: 37859-42-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR