DL-Methioninsulfoxid

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-4-(methylsulphinyl)butanoic acid

003117-5G 003117-25G
FLUO003117-5GEA 79.2 EUR
FLUO003117-5G FLUO003117-25G
DL-Methioninsulfoxid
DL-Methioninsulfoxid
Formel: C₅H₁₁NO₃S
Molecular Weight: 165,21 g/mol
MDL: MFCD00002620
CAS-Nummer: 62697-73-8
EINECS: 263-700-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR