2-Chlorethyl-ethylsulfid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

242640-25G 242640-5G
SIAL242640-25GEA 155 EUR
SIAL242640-25G SIAL242640-5G
2-Chlorethyl-ethylsulfid, Sigma-Aldrich®
2-Chlorethyl-ethylsulfid
Formel: C₄H₉ClS
MDL: MFCD00000979
CAS-Nummer: 693-07-2
EINECS: 211-742-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR