2-Chlorethyl-methylsulfid
Lieferant: FLUOROCHEM
Synonyme:
(2-Chloroethyl)(methyl)sulphane
003088-5G
003088-25G
FLUO003088-5GEA
0
EUR
FLUO003088-5G
FLUO003088-25G
2-Chlorethyl-methylsulfid
2-Chlorethyl-methylsulfid
Formel:
C₃H₇ClS Molecular Weight: 110,61 g/mol |
MDL:
MFCD00000978 CAS-Nummer: 542-81-4 EINECS: 208-828-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...