1,1,1-Trifluoracetoncyanhydrin 95%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
1,1,1-Trifluoroacetone Cyanohydrin, 2-Cyano-1,1,1-trifluoro-2-propanol, 2-Hydroxy-2-(trifluoromethyl)propionitrile
L11318.06
L11318.14
L11318.06EA
127
EUR
L11318.06
L11318.14
1,1,1-Trifluoracetoncyanhydrin 95%
1,1,1-Trifluoracetoncyanhydrin
Purity: 95%
CAS No.: 335-08-0
Molecular Formula: C4H4F3NO
Formula Weight: 139.08
Storage Temperature: -30°C to -10°C
Flash Point: 60°(140°F)
Refractive Index: 1.3380
Boiling Point: 68-69°/40mm
Beilstein Reference: 1758723
UN No.: 2929
MDL No.: MFCD00040885
Formel:
C₄H₄F₃NO Molecular Weight: 139.08 g/mol Siedepunkt: 68…69 °C (40 mmHg) Dichte: 1.276 Flammpunkt: 60 °C (140 °F) |
MDL:
MFCD00040885 CAS-Nummer: 335-08-0 UN: 2929 ADR: 6.1 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...