2-(Trifluormethyl)acrylsäure ≥98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 2-(Trifluoromethyl)propenoic acid

B22443.03 B22443.06 B22443.14
B22443.03EA 41 EUR
B22443.03 B22443.06 B22443.14
2-(Trifluormethyl)acrylsäure ≥98%
2-(Trifluormethyl)acrylsäure

Purity: 98% CAS No.: 381-98-6 Molecular Formula: C4H3F3O2 Formula Weight: 140.06 Flash Point: 82°(180°F) Melting Point: 49-51° UN No.: 3261 MDL No.: MFCD00042424

Formel: C₄H₃F₃O₂
Molecular Weight: 140,06 g/mol
Schmelzpunkt: 49…51 °C
Flammpunkt: 82 °C (180 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00042424
CAS-Nummer: 381-98-6
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR