2-(Trifluormethyl)acrylsäure

Lieferant: FLUOROCHEM

Synonyme: 2-(Trifluoromethyl)propenoic acid

007656-1G 007656-5G
FLUO007656-1GEA 30.6 EUR
FLUO007656-1G FLUO007656-5G
2-(Trifluormethyl)acrylsäure
2-(Trifluormethyl)acrylsäure
Formel: C₄H₃F₃O₂
Molecular Weight: 140,06 g/mol
MDL: MFCD00042424
CAS-Nummer: 381-98-6
UN: 3261
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR