2-(Ethoxycarbonyl)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

010794-5G 010794-1G
FLUO010794-5GEA 155 EUR
FLUO010794-5G FLUO010794-1G
2-(Ethoxycarbonyl)phenylboronsäurepinakolester
2-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
MDL: MFCD03453660
CAS-Nummer: 269409-99-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR