2-(Ethoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Ethoxycarbonylphenylboronic acid pinacol ester, 2-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, Ethyl 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

M-Clarity™ Qualitätsstufe = MQ100

570168-1G 570168-5G
SIAL570168-1GEA 102 EUR
SIAL570168-1G SIAL570168-5G
2-(Ethoxycarbonyl)phenylboronsäurepinakolester, Sigma-Aldrich®
2-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,136 g/mol
MDL: MFCD03453660
CAS-Nummer: 269409-99-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR