3-(Ethoxycarbonyl)phenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 3-Ethoxycarbonylphenylboronic acid, pinacol ester, Ethyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate
010795-5G
010795-1G
FLUO010795-5GEA
97.4
EUR
FLUO010795-5G
FLUO010795-1G
3-(Ethoxycarbonyl)phenylboronsäurepinakolester
3-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₅H₂₁BO₄ Molecular Weight: 276.136 g/mol |
MDL:
MFCD03093896 CAS-Nummer: 269410-00-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...