4-(Ethoxycarbonyl)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

PN-2113-1G PN-2113-5G
COBBPN-2113-1GEA 81.7 EUR
COBBPN-2113-1G COBBPN-2113-5G
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
Molecular Weight: 276,136 g/mol
MDL: MFCD03453659
CAS-Nummer: 195062-62-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR