4-(Ethoxycarbonyl)phenylboronsäurepinakolester

Lieferant: FLUOROCHEM

Synonyme: Ethyl 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate, 4-(Ethoxycarbonyl)benzeneboronic acid pinacol ester, Ethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoate

010796-5G 010796-1G
FLUO010796-5GEA 291 EUR
FLUO010796-5G FLUO010796-1G
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
4-(Ethoxycarbonyl)phenylboronsäurepinakolester
Formel: C₁₅H₂₁BO₄
MDL: MFCD03453659
CAS-Nummer: 195062-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR