3-Carboxyphenylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
367570010
367570050
ACRO367570010EA
32.5
EUR
ACRO367570010
ACRO367570050
3-Carboxyphenylboronsäurepinakolester 97%
3-Carboxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molecular Weight: 248.083 g/mol Schmelzpunkt: 206…211 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD03411930 CAS-Nummer: 269409-73-6 |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 206 to 211 °C |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...