3-Carboxyphenylboronsäurepinakolester
Lieferant: FLUOROCHEM
Synonyme:
3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid
010828-2G
010828-5G
010828-25G
FLUO010828-2GEA
0
EUR
FLUO010828-2G
FLUO010828-5G
FLUO010828-25G
3-Carboxyphenylboronsäurepinakolester
3-Carboxyphenylboronsäurepinakolester
Formel:
C₁₃H₁₇BO₄ Molecular Weight: 248.083 g/mol |
MDL:
MFCD03411930 CAS-Nummer: 269409-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...