4-Carboxyphenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 4-Carboxybenzeneboronic acid pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

BB-2105-25G BB-2105-5G BB-2105-1G
COBBBB-2105-25GEA 130 EUR
COBBBB-2105-25G COBBBB-2105-5G COBBBB-2105-1G
4-Carboxyphenylboronsäurepinakolester
4-Carboxyphenylboronsäurepinakolester
Formel: C₁₃H₁₇BO₄
Molecular Weight: 248,08 g/mol
MDL: MFCD01863710
CAS-Nummer: 180516-87-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR