4-Carboxy-2-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

PN-2451-5G PN-2451-25G PN-2451-1G
COBBPN-2451-5GEA 451 EUR
COBBPN-2451-5G COBBPN-2451-25G COBBPN-2451-1G
4-Carboxy-2-nitrophenylboronsäurepinakolester
4-Carboxy-2-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₆
MDL: MFCD12546615
CAS-Nummer: 1218791-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR