3-Carboxy-5-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 3-Carboxy-5-nitrobenzeneboronic acid pinacol ester, 3-Nitro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid, 3-Carboxy-5-nitrophenylboronic acid, pinacol ester

BB-2005-5G BB-2005-1G
COBBBB-2005-5GEA 375 EUR
COBBBB-2005-5G COBBBB-2005-1G
3-Carboxy-5-nitrophenylboronsäurepinakolester
3-Carboxy-5-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₆BNO₆
Molecular Weight: 293,08 g/mol
MDL: MFCD03411932
CAS-Nummer: 377780-80-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR