3-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
BB-3856-5G
BB-3856-1G
COBBBB-3856-5GEA
149
EUR
COBBBB-3856-5G
COBBBB-3856-1G
3-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
3-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
Formel:
C₁₅H₁₆BNO₃ |
MDL:
MFCD09027261 CAS-Nummer: 957060-98-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...