3-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3859-5G BB-3859-1G
COBBBB-3859-5GEA 303 EUR
COBBBB-3859-5G COBBBB-3859-1G
3-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure
3-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure
Formel: C₁₅H₁₆BNO₃
MDL: MFCD09027263
CAS-Nummer: 957061-00-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR