4-(2,3-Dimethylphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3205-5G BB-3205-1G
COBBBB-3205-5GEA 375 EUR
COBBBB-3205-5G COBBBB-3205-1G
4-(2,3-Dimethylphenylcarbamoyl)phenylboronsäure
4-(2,3-Dimethylphenylcarbamoyl)phenylboronsäure
Formel: C₁₅H₁₆BNO₃
Molecular Weight: 269.1 g/mol
MDL: MFCD09027210
CAS-Nummer: 913835-36-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR