4-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

BB-3207-5G BB-3207-1G
COBBBB-3207-5GEA 149 EUR
COBBBB-3207-5G COBBBB-3207-1G
4-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
4-(2,4-Dimethylphenylcarbamoyl)phenylboronsäure
Formel: C₁₅H₁₆BNO₃
MDL: MFCD09027212
CAS-Nummer: 913835-38-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR