4-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(4-((2,5-Dimethylphenyl)carbamoyl)phenyl)boronic acid
BB-3209-5G
BB-3209-1G
COBBBB-3209-5GEA
523
EUR
COBBBB-3209-5G
COBBBB-3209-1G
4-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure
4-(2,5-Dimethylphenylcarbamoyl)phenylboronsäure
Formel:
C₁₅H₁₆BNO₃ Molecular Weight: 269.1 g/mol |
MDL:
MFCD09027214 CAS-Nummer: 913835-40-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...